missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals

Product Code. 11410191
Change view
Click to view available options
Quantity:
250 mL
500 mL
1000 mL
Unit Size:
1 L
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410191 1000 mL 1 L
11319027 250 mL 250mL
11400191 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11410191 Supplier Thermo Scientific Alfa Aesar Supplier No. 038746.AU

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
SMILES F

Specifications

Color Colorless
Boiling Point 112°C
Density 1.16g/mL
Odor Acrid
Quantity 1000 mL
Physical Form Liquid, Double Sub-Boiling Quartz Distillation
UN Number UN1790
Merck Index 14,4790
IUPAC Name hydrogen fluoride
Formula Weight 20.01
Percent Purity 47 to 51%
Grade Environmental Plus
Chemical Name or Material Hydrofluoric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.