missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, 99.99% (metals basis) 40% min, Thermo Scientific Chemicals

Product Code. p-7048597
Click to view available options
Quantity:
25 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
25mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11311588

Brand: Thermo Scientific Alfa Aesar 010989.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hydrofluoric acid is used in the glass etching, electronic and chemical industries. Hydrogen fluoride is mainly used in the production of organofluorine compounds, aluminum and chlorofluorocarbons. Hydrogen fluoride is also used for separating uranium isotopes, as a catalyst in the petroleum industry, and in stainless steel pickling. Hydrofluoric acid is used as a pickling agent and used in the semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Color Colorless
Boiling Point 112°C
Density 1.16 g/mL
Odor Acrid
Sensitivity Hygroscopic
Quantity 25 mL
Physical Form Liquid
UN Number UN1790
Merck Index 14,4790
Solubility Information Strongly miscible with water and alcohol. Slightly miscible with ether and miscible with many organic solvents.
Formula Weight 20.01
Percent Purity 99.99%
Assay (metals basis) 40% min
Chemical Name or Material Hydrofluoric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.