missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, 48%, Honeywell™ Fluka™
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
pH | Acidic |
Color | Colorless |
Melting Point | -35°C |
Boiling Point | ∼105°C |
Density | 1.15g/mL at 25°C |
Odor | Stinging |
Packaging | Poly bottle |
Vapor Pressure | 25mmHg (20°C) |
Quantity | 500 mL |
Physical Form | Liquid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'