missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 48%, Honeywell™ Fluka™

Product Code. 15608460 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Poly bottle
Unit Size:
25mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15608460 25 mL Poly bottle 25mL
15636720 500 mL Poly bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15608460 Supplier Honeywell-Fluka Supplier No. 69506825ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 48%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

pH Acidic
Color Colorless
Melting Point -35°C
Boiling Point ∼105°C
Density 1.15g/mL at 25°C
Odor Stinging
Packaging Poly bottle
Vapor Pressure 25mmHg (20°C)
Quantity 25 mL
Physical Form Liquid
UN Number UN1790B
Formula Weight 20.01g/mol
Percent Purity 48%
Chemical Name or Material Hydrofluoric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.