missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 48%, for Electronic Use, Fisher Chemical

Product Code. 10676832 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10676832 2.5 L HDPE plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10676832 Supplier Fisher Chemical Supplier No. H/1425/17

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

pH 1
Color Colorless
CAS 7664-39-3
Melting Point -35°C
Boiling Point 105°C
Packaging HDPE plastic bottle
Quantity 2.5 L
Molecular Formula FH
Physical Form Liquid
MDL Number MFCD00011346
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES F
IUPAC Name hydrogen fluoride
Molecular Weight (g/mol) 20.01
PubChem CID 14917
ChEBI CHEBI:29228
Formula Weight 20g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrofluoric acid, 48 to 51% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.