missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, 46% J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Color | Colorless |
| CAS | 7664-39-3 |
| Melting Point | 35°C |
| Boiling Point | 105°C |
| Density | 1.2g/cm3 |
| Odor | Pungent |
| Assay Percent Range | 47 to 51% |
| Quantity | 500 mL |
| Molecular Formula | FH |
| Physical Form | Liquid |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction