missing translation for 'onlineSavingsMsg'
Learn More

Hydrodolasetron, TRC

Product Code. 30390376
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390376 50 mg 50mg
30397566 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390376 Supplier Toronto Research Chemicals Supplier No. TRCH71475550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127951-99-9
Molecular Formula C19 H22 N2 O3
Molecular Weight (g/mol) 326.39
Synonym 2,6-Methano-2H-quinolizine, 1H-indole-3-carboxylic acid deriv., 1H-Indole-3-carboxylic Acid Octahydro-3-hydroxy-2,6-methano-2H-quinolizin-8-yl Ester, MDL 74156, Reduced Dolasetron
SMILES O[C@@H]1CN2[C@@H]3CC(CC2C[C@H]1C3)OC(=O)c4c[nH]c5ccccc45

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H22N2O3/c22-18-10-21-12-5-11(18)6-13(21)8-14(7-12)24-19(23)16-9-20-17-4-2-1-3-15(16)17/h1-4,9,11-14,18,20,22H,5-8,10H2/t11-,12+,13?,14?,18-/m1/s1
Formula Weight 326.163
Chemical Name or Material Hydrodolasetron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.