missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Hydrodex γ-TBDAc GC Capillary Column

Brand:  Macherey-Nagel™ 723387.50

 View more versions of this product

Product Code. 12412770

  • £1650.00 / Each
Estimated Shipment: 31-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

0.4 mm
0.25 mm
Enantiomer Separation
50 m
1 Each
GC Capillary Column
Hydrodex γ-TBDAc
220°C to 240°C
Hydrodex γ-TBDAc
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers