missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Hydrocortisone Stock Solution

Product Code. 17109921
Encompass
Change view
Click to view available options
Quantity:
3 mL
10 x 3 mL
Unit Size:
3mL
Pack of 10
Product Code. Quantity unitSize
17109921 10 x 3 mL Pack of 10
17199911 3 mL 3mL
2 options
This item is not returnable. View return policy

Product Code. 17109921

Brand: STEMCELL Technologies 07926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrocortisone Stock Solution (200X) for use with PneumaCult™-ALI, MammoCult™, EpiCult™, 10 pack

  • Hydrocortisone Stock Solution is recommended for use as a cell culture supplement and for the preparation of PneumaCult™, MammoCult™, and EpiCult™ media when culturing airway epithelial cells, mammary primary cells, or tumor cell lines
  • Hydrocortisone Stock Solution is provided at a concentration of 96 μg/mL, which is ideal as a 200X stock solution for use with PneumaCult™-ALI (Catalog #05001), MammoCult™ (Catalog #05620), EpiCult™-B (Human; Catalog #05601), and EpiCult™-C (Catalog #05630) media
  • It may also be used for preparing PneumaCult™-Ex Medium (Catalog #05008) or PneumaCult™-Ex Plus Medium (Catalog #05040)

Specifications

Cell Type Airway Cells, Cancer Cells and Cell Lines, Mammary Cells
For Use With (Application) Cell Culture
Quantity 10 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.