missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone Sodium Succinate, Mikromol™

Product Code. 30050459
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050459 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050459 Supplier Mikromol Supplier No. MM3100.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hydrocortisone Sodium Succinate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocortisone Sodium Succinate
CAS 125-04-2
Analyte or Component Names Hydrocortisone Sodium Succinate
Shipping Condition Ice pack (-20°C)
Molecular Formula C25 H33 O8 . Na
Certifications/Compliance ISO 17034
Synonym Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, sodium salt (1:1), (11β)-, Cortisol, 21-(hydrogen succinate), monosodium salt (8CI), Cortisol, hydrogen succinate, sodium salt (7CI), Cortisol, succinate, sodium salt (6CI), Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, monosodium salt, (11β)- (9CI), A-HydroCort, Buccalsone, Corlan, Cortisol 21-sodium succinate, Cortisol hemisuccinate sodium salt, Cortisol sodium hemisuccinate, Cortisol sodium succinate, EF-Cortelan Soluble, Emi-Corlin, Flebocortid, Hydrocortisone 21-hemisuccinate sodium salt, Hydrocortisone 21-sodium succinate, Hydrocortisone 21-sodium succinide, Hydrocortisone 21-succinate sodium salt, Hydrocortisone hemisuccinate sodium salt, Hydrocortisone sodium 21-hemisuccinate, Hydrocortisone sodium hemisuccinate, Hydrocortisone sodium succinate, Intracort, Nordicort, Oralsone, Sodium 17-hydroxycorticosterone 21-succinate, Sodium hydrocortisone 21-hemisuccinate, Sodium hydrocortisone 21-succinate, Sodium hydrocortisone succinate, Solu-Cortef, Solu-Glyc, U 4905
InChI Formula InChI=1S/C25H34O8.Na/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30);/q;+1/p-1/t16-,17-,18-,22+,23-,24-,25-;/m0./s1
SMILES [Na+].C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(=O)[O-])[C@@]4(C)C[C@H](O)[C@H]23
IUPAC Name sodium;4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoate
Molecular Weight (g/mol) 484.51
Quantity 250 mg
Formula Weight 484.2073 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.