missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone, MedChemExpress

Product Code. 16442539
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442539 1 g 1g
16411448 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442539 Supplier Medchem Express Supplier No. HYN05831g
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hydrocortisone (Cortisol) is a steroid hormone or glucocorticoid secreted by the adrenal cortex.

Hydrocortisone (Cortisol) is a steroid hormone or glucocorticoid secreted by the adrenal cortex.

Specifications

Chemical Name or Material Hydrocortisone
CAS 50-23-7
Color White
Purity Grade Notes Research
Molecular Formula C21H30O5
Quantity 1 g
Synonym Cortisol
Solubility Information DMSO : ≥ 31 mg/mL (85.53 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]1([C@@]2(O)C(CO)=O)[C@](CC2)([H])[C@@](CCC3=CC4=O)([H])[C@]([C@]3(CC4)C)([H])[C@@H](O)C1
Molecular Weight (g/mol) 362.46
Formula Weight 362.46
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.