missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone Hemisuccinate Sodium Salt, TRC

Product Code. 30012715
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012715 1 g 1g
30004481 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012715 Supplier Toronto Research Chemicals Supplier No. TRCH7146801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-04-2
Molecular Formula C25 H33 O8 . Na
Molecular Weight (g/mol) 484.51
Synonym Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, sodium salt (1:1), (11β)-, Cortisol, 21-(hydrogen succinate), monosodium salt (8CI), Cortisol, hydrogen succinate, sodium salt (7CI), Cortisol, succinate, sodium salt (6CI), Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, monosodium salt, (11β)- (9CI), A-HydroCort, Buccalsone, Corlan, Cortisol 21-sodium succinate, Cortisol hemisuccinate sodium salt, Cortisol sodium hemisuccinate, Cortisol sodium succinate, EF-Cortelan Soluble, Emi-Corlin, Flebocortid, Hydrocortisone 21-hemisuccinate sodium salt, Hydrocortisone 21-sodium succinate, Hydrocortisone 21-sodium succinide, Hydrocortisone 21-succinate sodium salt, Hydrocortisone hemisuccinate sodium salt, Hydrocortisone sodium 21-hemisuccinate, Hydrocortisone sodium hemisuccinate, Hydrocortisone sodium succinate, Intracort, Nordicort, Oralsone, Sodium 17-hydroxycorticosterone 21-succinate, Sodium hydrocortisone 21-hemisuccinate, Sodium hydrocortisone 21-succinate, Sodium hydrocortisone succinate, Solu-Cortef, Solu-Glyc, U 4905
IUPAC Name sodium;4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoate
SMILES [Na+].C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(=O)[O-])[C@@]4(C)C[C@H](O)[C@H]23

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H34O8.Na/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30);/q;+1/p-1/t16-,17-,18-,22+,23-,24-,25-;/m0./s1
Formula Weight 484.2073
Chemical Name or Material Hydrocortisone Sodium Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.