missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone (Cortisol), Mikromol™

Product Code. 18011610
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18011610 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18011610 Supplier Mikromol Supplier No. MM0185.000250
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hydrocortisone (Cortisol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocortisone
CAS 50-23-7
Analyte or Component Names Hydrocortisone
Shipping Condition Room Temperature
Molecular Formula C21 H30 O5
Certifications/Compliance ISO 17034
Synonym Hydrocortisone, Cortisol, 11beta,17,21-Trihydroxy-pregn-4-ene-3,20-dione, Hydrocortisone Hydrogen Succinate Imp. A (EP), Prednisolone Imp. A (EP), Hydrocortisone Acetate Imp. A (Pharmeuropa), 17-Hydroxycorticosterone, Acticort, Aeroseb HC, Ala-Cort, Anflam, Anti-inflammatory hormone, CCN 90306A, CaldeCort Spray, Cetacort, Cobadex, Cort-Dome, Cortanal, Cortef, Cortenema, Corticreme, Cortifan, Cortiment, Cortispray, Cortonema, Cortril, Dermacort, Dermocortal, Dermolate, Dihydrocostisone, Dioderm, Domolene-HC, Efcorbin, Efcortelan, Eldecort, Epiderm H, Esiderm H, Evacort, Ficortril, Genacort, HC, Heb-Cort, Hidro-Colisona, Hycort, Hycortol, Hycortole, Hydracort, Hydrasson, Hydro-Adreson, Hydrocort, Hydrocortal, Hydrocorticosterone, Hydrocortisyl, Hydrocortone, Hydroskin, Hytone, Hytone lotion, Incortin-H, Kendall's compound F, Lacticare HC, Medicort, Mildison, NSC 10483, Nutracort, Optef, Otosone-F, Penecort, Permicort, Polcort H, Reichstein's substance M, Scheroson F, Signef, Synacort, Tarcortin, Texacort, Texacort lotion 25, Timocort, Topicort, Traumaide, U 1851, Zenoxone, 11beta,17,21-Trihydroxyprogesterone, 11beta,17alpha,21-Trihydroxypregn-4-ene-3,20-dione, 11beta-Hydroxycortisone, 17alpha-Hydroxycorticosterone, 4-Pregnene-11beta,17alpha,21-triol-3,20-dione, Hydrocortisone, Hydrocortisone Acetate Impurity A, Hydrocortisone Hydrogen Succinate Impurity A, Prednisolone Impurity A
InChI Formula InChI=1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 362.46
Quantity 250 mg
Formula Weight 362.2093 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.