missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone (Cortisol), LoGiCal

Product Code. 30459926
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459926 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459926 Supplier LoGiCal Supplier No. LGCFOR0185.00
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocortisone
CAS 50-23-7
Analyte or Component Names Hydrocortisone
Shipping Condition Room Temperature
Molecular Formula C21 H30 O5
Certifications/Compliance ISO 9001
Synonym Hydrocortisone, Cortisol, 11beta,17,21-Trihydroxy-pregn-4-ene-3,20-dione, Hydrocortisone Hydrogen Succinate Imp. A (EP), Prednisolone Imp. A (EP), Hydrocortisone Acetate Imp. A (Pharmeuropa), 17-Hydroxycorticosterone, Acticort, Aeroseb HC, Ala-Cort, Anflam, Anti-inflammatory hormone, CCN 90306A, CaldeCort Spray, Cetacort, Cobadex, Cort-Dome, Cortanal, Cortef, Cortenema, Corticreme, Cortifan, Cortiment, Cortispray, Cortonema, Cortril, Dermacort, Dermocortal, Dermolate, Dihydrocostisone, Dioderm, Domolene-HC, Efcorbin, Efcortelan, Eldecort, Epiderm H, Esiderm H, Evacort, Ficortril, Genacort, HC, Heb-Cort, Hidro-Colisona, Hycort, Hycortol, Hycortole, Hydracort, Hydrasson, Hydro-Adreson, Hydrocort, Hydrocortal, Hydrocorticosterone, Hydrocortisyl, Hydrocortone, Hydroskin, Hytone, Hytone lotion, Incortin-H, Kendall's compound F, Lacticare HC, Medicort, Mildison, NSC 10483, Nutracort, Optef, Otosone-F, Penecort, Permicort, Polcort H, Reichstein's substance M, Scheroson F, Signef, Synacort, Tarcortin, Texacort, Texacort lotion 25, Timocort, Topicort, Traumaide, U 1851, Zenoxone, 11beta,17,21-Trihydroxyprogesterone, 11beta,17alpha,21-Trihydroxypregn-4-ene-3,20-dione, 11beta-Hydroxycortisone, 17alpha-Hydroxycorticosterone, 4-Pregnene-11beta,17alpha,21-triol-3,20-dione, Hydrocortisone, Hydrocortisone Acetate Impurity A, Hydrocortisone Hydrogen Succinate Impurity A, Prednisolone Impurity A
InChI Formula InChI=1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
Solution Type Neat
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 362.46
Quantity 10 mg
Formula Weight 362.2093 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.