missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone Butyrate, TRC

Product Code. 30405308
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405308 1 g 1g
30395456 10 g -
30393241 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405308 Supplier Toronto Research Chemicals Supplier No. TRCH7146051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13609-67-1
Molecular Formula C25 H36 O6
Molecular Weight (g/mol) 432.55
Synonym Cortisol, 17-butyrate (6CI,8CI), (11β)-11,21-Dihydroxy-17-(1-oxobutoxy)pregn-4-ene-3,20-dione, Butyric acid, 17-ester with cortisol (8CI), 4-Pregnene-11β,21-dihydroxy-17α-butyryloxy-3,20-dione, Alfason, Hydrocortisone 17-butyrate, Hydrocortisone 17α-butyrate, Hydrocortisone butyrate, Laticort, Locoid, Plancol
IUPAC Name [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate
SMILES CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H36O6/c1-4-5-21(30)31-25(20(29)14-26)11-9-18-17-7-6-15-12-16(27)8-10-23(15,2)22(17)19(28)13-24(18,25)3/h12,17-19,22,26,28H,4-11,13-14H2,1-3H3/t17-,18-,19-,22+,23-,24-,25-/m0/s1
Formula Weight 432.2512
Chemical Name or Material Hydrocortisone Butyrate (Hydrocortisone 17-Butyrate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.