missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocortisone Butyrate (Hydrocortisone 17-Butyrate), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Hydrocortisone Butyrate (Hydrocortisone 17-Butyrate) |
| CAS | 13609-67-1 |
| Analyte or Component Names | Hydrocortisone Butyrate (Hydrocortisone 17-Butyrate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H36 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Hydrocortisone 17-Butyrate, Cortisol, 17-butyrate (6CI,8CI), Butyric acid, 17-ester with cortisol (8CI), Alfason, Hydrocortisone butyrate, Laticort, Locoid, Plancol, (11beta)-11,21-Dihydroxy-17-(1-oxobutoxy)pregn-4-ene-3,20-dione, 4-Pregnene-11beta,21-dihydroxy-17alpha-butyryloxy-3,20-dione, Hydrocortisone 17-butyrate, Hydrocortisone 17alpha-butyrate |
| InChI Formula | InChI=1S/C25H36O6/c1-4-5-21(30)31-25(20(29)14-26)11-9-18-17-7-6-15-12-16(27)8-10-23(15,2)22(17)19(28)13-24(18,25)3/h12,17-19,22,26,28H,4-11,13-14H2,1-3H3/t17-,18-,19-,22+,23-,24-,25-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?