missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocortisone Acetate, Mikromol™
Description
Hydrocortisone Acetate
Specifications
Specifications
| Chemical Name or Material | Hydrocortisone Acetate |
| CAS | 50-03-3 |
| Analyte or Component Names | Hydrocortisone Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H32 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cortisone Acetate Imp. A (EP), Hydrocortisone Hydrogen Succinate Imp. B (EP), Hydrocortisone Imp. C (EP), Prednisolone Acetate Imp. A (EP), Hydrocortisone Acetate, Hydrocortisone 21-Acetate, (11beta)-21-(Acetyloxy)-11,17-dihydroxypregn-4-ene-3,20-dione, Cortisol acetate, Cortisol 21-acetate, 11beta,17alpha,21-Trihydroxypregn-4-ene-3,20-dione 21-acetate, 17-Hydroxycorticosterone 21-acetate, 17alpha-Hydroxycorticosterone acetate, 21-Acetoxy-11beta,17-dihydroxypregn-4-en-3,20-dione, 21-Acetoxy-11beta,17-dihydroxypregn-4-ene-3,20-dione, 21-Acetoxy-11beta,17alpha-dihydroxypregn-4-ene-3,20-dione, Abbocort, Anusol HC, Bambicort, Berlison F, Biocortar, CaldeCort, Chemysone, Colifoam, Colofoam, Cordes, Cortacream, Cortaid, Cortef acetate, Cortell, Corticaine, Cortifoam, Cortril acetate, Cortril acetate AS, Efcolin, HA, Hc 45, Hycortole acetate, Hydrin 2, Hydrocal, Hydrocort acetate, Hydrocortisat, Hydrocortisone 21-acetate, Hydrocortisone acetate, Hydrocortistab, Hydrocortone acetate, Hydroxycorticosterone acetate, Isopto-Hydrocortisone, Lanacort, Lenirit, NSC 741, Pabracort, Proctocort, Sigmacort, Sintotrat, Velopural, Wycort, Cortisone Acetate Impurity A, Hydrocortisone Hydrogen Succinate Impurity B, Hydrocortisone Impurity C, Prednisolone Acetate Impurity A |
| InChI Formula | InChI=1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1 |
| SMILES | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C |
| IUPAC Name | [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?