missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocortisone acetate, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Hydrocortisone acetate is a corticosteroid, used to decrease swelling, itching, and pain that is caused by minor skin irritations or by hemorrhoids.
Specifications
Specifications
| Chemical Name or Material | Hydrocortisone acetate |
| CAS | 50-03-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H32O6 |
| Quantity | 100 mg |
| Synonym | Hydrocortisone 21-acetate Cortisol 21-acetate |
| Solubility Information | DMSO : ≥ 38 mg/mL (93.94 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | C[C@@]12[C@](C(COC(C)=O)=O)(O)CC[C@@]1([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])[C@@H](O)C2)=O |
| Molecular Weight (g/mol) | 404.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction