missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocortisone Acetate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Hydrocortisone Acetate |
| CAS | 50-03-3 |
| Analyte or Component Names | Hydrocortisone Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H32 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Cortisone Acetate Imp. A (EP), Hydrocortisone Hydrogen Succinate Imp. B (EP), Hydrocortisone Imp. C (EP), Prednisolone Acetate Imp. A (EP), Hydrocortisone Acetate, Hydrocortisone 21-Acetate, (11beta)-21-(Acetyloxy)-11,17-dihydroxypregn-4-ene-3,20-dione, Cortisol acetate, Cortisol 21-acetate, 11beta,17alpha,21-Trihydroxypregn-4-ene-3,20-dione 21-acetate, 17-Hydroxycorticosterone 21-acetate, 17alpha-Hydroxycorticosterone acetate, 21-Acetoxy-11beta,17-dihydroxypregn-4-en-3,20-dione, 21-Acetoxy-11beta,17-dihydroxypregn-4-ene-3,20-dione, 21-Acetoxy-11beta,17alpha-dihydroxypregn-4-ene-3,20-dione, Abbocort, Anusol HC, Bambicort, Berlison F, Biocortar, CaldeCort, Chemysone, Colifoam, Colofoam, Cordes, Cortacream, Cortaid, Cortef acetate, Cortell, Corticaine, Cortifoam, Cortril acetate, Cortril acetate AS, Efcolin, HA, Hc 45, Hycortole acetate, Hydrin 2, Hydrocal, Hydrocort acetate, Hydrocortisat, Hydrocortisone 21-acetate, Hydrocortisone acetate, Hydrocortistab, Hydrocortone acetate, Hydroxycorticosterone acetate, Isopto-Hydrocortisone, Lanacort, Lenirit, NSC 741, Pabracort, Proctocort, Sigmacort, Sintotrat, Velopural, Wycort, Cortisone Acetate Impurity A, Hydrocortisone Hydrogen Succinate Impurity B, Hydrocortisone Impurity C, Prednisolone Acetate Impurity A |
| InChI Formula | InChI=1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction