missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone, ≥97%, MP Biomedicals™

Product Code. 11354469 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354469 100 g 100g
11314469 1 g 1g
11324469 5 g 5g
11334469 10 g 10g
11344469 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11354469 Supplier MP Biomedicals Supplier No. 0210199680
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-23-7
Molecular Formula C21H30O5
Molecular Weight (g/mol) 362.466
MDL Number MFCD00011654
InChI Key JYGXADMDTFJGBT-VWUMJDOOSA-N
Synonym hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
PubChem CID 5754
ChEBI CHEBI:17650
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O

Specifications

Color White
Assay Percent Range ≥97%
Quantity 100 g
Formula Weight 362.5
Physical Form Crystalline Powder
Chemical Name or Material Hydrocortisone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.