missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Hydrocortisone
Description
- Hydrocortisone is a glucocorticoid hormone with anti-inflammatory and immunosuppressive effects (Kang et al; Langhoff & Ladefoged)
- It is also commonly used as a supplement for endothelial, epithelial, mesenchymal, or oligodendrocyte cell culture media, as it supports growth and differentiation
- This product is supplied as the sodium salt of hydrocortisone 21-hemisuccinate, which is a more water-soluble form of hydrocortisone
- Hydrocortisone is required as a supplement in MyeloCult™ H5100 (Human; Catalog #05100) and MyeloCult™ M5300 (Mouse; Catalog #05300), for long-term cultures and the long-term culture-initiating cell (LTC-IC) assay
Specifications
Specifications
| CAS | 125-04-2 |
| Synonym | Cortisol 21-hemisuccinate; Hydrocortisone sodium succinate; ST51037292; U-4905 |
| Quantity | 100 mg |
| Molecular Formula | C25H33O8 · Na |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?