missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 3-(O-Carboxymethyl)oxime (E/Z Mixture), TRC

Product Code. 30408218
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408218 1 g -
30393240 100 mg -
30395600 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408218 Supplier Toronto Research Chemicals Supplier No. TRCH7165681G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43188-86-9
Molecular Formula C23 H33 N O7
Molecular Weight (g/mol) 435.511
Synonym 2-[[[(11beta)-11,17,21-Trihydroxy-20-oxopregn-4-en-3-ylidene]amino]oxy]acetic Acid, Cortisol 3-(O-Carboxymethyl)oxime, Cortisol 3-(Carboxymethyloxime), [[[(11beta)-11,17,21-Trihydroxy-20-oxopregn-4-en-3-ylidene]amino]oxy]acetic Acid
IUPAC Name 2-[(E)-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-ylidene]amino]oxyacetic acid
SMILES C[C@]12CC\C(=N/OCC(=O)O)\C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23

Specifications

Quantity 1 g
InChI Formula InChI=1S/C23H33NO7/c1-21-7-5-14(24-31-12-19(28)29)9-13(21)3-4-15-16-6-8-23(30,18(27)11-25)22(16,2)10-17(26)20(15)21/h9,15-17,20,25-26,30H,3-8,10-12H2,1-2H3,(H,28,29)/b24-14+/t15-,16-,17-,20+,21-,22-,23-/m0/s1
Formula Weight 435.226
Chemical Name or Material Hydrocortisone 3-(O-Carboxymethyl)oxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.