missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hydrocortisone-21-Acetate, MP Biomedicals™

Product Code. 11425975 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11425975

Brand: MP Biomedicals™ 0210199525

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibits most inflammatory and immune responses of allergic, bacterial, traumatic or anaphylactic origin. Hydrocortisone-21-Acetate, MP Biomedicals is an immunosuppressant drug that acts by inhibiting migration of leukocytes and phagocytes and decreasing

  • Classified as a corticosteroid (e.g. betamethasone, dexamethasone, fluorometholone, hydrocortisone acetate, loteprednol etabonate, prednisolone, rimexolone, triamcinolone)
  • They are sometimes combined with an antibiotic drug (e.g. betamethasone combined with neomycin or sulfacetamide, dexamethasone combined with neomycin or polymyxin B)
  • Primary functions are to increase blood sugar through gluconeogenesis and to suppresses inflammatory and immune responses
TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Store at Room Temperature; desiccate. Protect from light.
Form Powder
Quantity 25 g
Regulatory Status RUO
compliance-icons
Product Identifier
  • Hydrocortisone-21-acetate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocortisone-21-acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.