missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocortisone-21-Acetate, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| IUPAC Name | 2-[(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate |
| Formula Weight | 404.5 |
| Physical Form | Solid |
| Chemical Name or Material | Hydrocortisone-21-Acetate |
Safety and Handling
Signal Word
- Warning
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction