missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone-21-Acetate, MP Biomedicals™

Product Code. 11394459 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 11394459

Brand: MP Biomedicals 0210199505

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-03-3
Molecular Formula C23H32O6
Molecular Weight (g/mol) 404.50
MDL Number MFCD00037714
InChI Key ALEXXDVDDISNDU-JZYPGELDSA-N
Synonym hydrocortisone acetate, cortell, cortisol 21-acetate, cortisol acetate, cortacream, hydrocortisat, abbocort, bambicort, biocortar, chemysone
PubChem CID 5744
ChEBI CHEBI:17609
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

Color White
Quantity 5 g
IUPAC Name 2-[(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Formula Weight 404.5
Physical Form Solid
Chemical Name or Material Hydrocortisone-21-Acetate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.