missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 21-Acetate-D3, TRC

Product Code. 30394639
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394639 50 mg -
30404501 250 mg -
30394637 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394639 Supplier Toronto Research Chemicals Supplier No. TRCH71462250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C23H29D3O6
Molecular Weight (g/mol) 407.52
SMILES [H][C@]1([C@@H](O)C[C@@]2([C@](C(COC(C([2H])([2H])[2H])=O)=O)(O)CC3)C)[C@]([H])([C@@]23[H])CCC4=CC(CC[C@@]41C)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1/i1D3
Chemical Name or Material Hydrocortisone 21-Acetate-D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.