missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 17-Propionate 21-Acetate, TRC

Product Code. 30394340
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394340 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394340 Supplier Toronto Research Chemicals Supplier No. TRCH71462825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocortisone 17-Propionate 21-Acetate
CAS 74050-20-7
Molecular Formula C26 H36 O7
Quantity 25 mg
Synonym Pregn-4-ene-3,20-dione, 21-(acetyloxy)-11-hydroxy-17-(1-oxopropoxy)-, (11β)-, (11β)-21-(Acetyloxy)-11-hydroxy-17-(1-oxopropoxy)pregn-4-ene-3,20-dione, Cortisol 17-propionate 21-acetate, Hydrocortisone 17-propionate 21-acetate, Hydrocortisone aceponate, Retef, Hydrocortisone 17-Propionate 21-Acetate
InChI Formula InChI=1S/C26H36O7/c1-5-22(31)33-26(21(30)14-32-15(2)27)11-9-19-18-7-6-16-12-17(28)8-10-24(16,3)23(18)20(29)13-25(19,26)4/h12,18-20,23,29H,5-11,13-14H2,1-4H3/t18-,19-,20-,23+,24-,25-,26-/m0/s1
SMILES CCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)C
IUPAC Name [(8S,9S,10R,11S,13S,14S,17R)-17-(2-acetyloxyacetyl)-11-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 460.56
Formula Weight 460.2461
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.