missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 17-Butyrate, Mikromol™

Product Code. 17952576
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952576 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17952576 Supplier Mikromol Supplier No. MM0188.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hydrocortisone 17-Butyrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocortisone Butyrate (Hydrocortisone 17-Butyrate)
CAS 13609-67-1
Analyte or Component Names Hydrocortisone Butyrate (Hydrocortisone 17-Butyrate)
Shipping Condition Room Temperature
Molecular Formula C25 H36 O6
Certifications/Compliance ISO 17034
Synonym Hydrocortisone 17-Butyrate, Cortisol, 17-butyrate (6CI,8CI), Butyric acid, 17-ester with cortisol (8CI), Alfason, Hydrocortisone butyrate, Laticort, Locoid, Plancol, (11beta)-11,21-Dihydroxy-17-(1-oxobutoxy)pregn-4-ene-3,20-dione, 4-Pregnene-11beta,21-dihydroxy-17alpha-butyryloxy-3,20-dione, Hydrocortisone 17-butyrate, Hydrocortisone 17alpha-butyrate
InChI Formula InChI=1S/C25H36O6/c1-4-5-21(30)31-25(20(29)14-26)11-9-18-17-7-6-15-12-16(27)8-10-23(15,2)22(17)19(28)13-24(18,25)3/h12,17-19,22,26,28H,4-11,13-14H2,1-3H3/t17-,18-,19-,22+,23-,24-,25-/m0/s1
SMILES CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO
IUPAC Name [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate
Molecular Weight (g/mol) 432.55
Quantity 500 mg
Formula Weight 432.2512 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.