missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 17-butyrate, MedChemExpress

Product Code. 16401456
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401456 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401456 Supplier Medchem Express Supplier No. HYB0983

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Hydrocortisone 17-butyrate is an adrenocortico hormone

Hydrocortisone 17-butyrate is an adrenocortico hormone

Specifications

Chemical Name or Material Hydrocortisone 17-butyrate
CAS 13609-67-1
Color White
Purity Grade Notes Research
Molecular Formula C25H36O6
Quantity 200 mg
Synonym Cortisol 17-butyrate Hydrocortisone butyrate
Solubility Information DMSO : 100 mg/mL (231.19 mM; Need ultrasonic)
SMILES C[C@@]12[C@](C(CO)=O)(OC(CCC)=O)CC[C@@]1([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])[C@@H](O)C2)=O
Molecular Weight (g/mol) 432.55
Formula Weight 432.55
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.