missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocortisone 17-Butyrate 21-Propionate, TRC
Specifications
Specifications
| Chemical Name or Material | Hydrocortisone 17-Butyrate 21-Propionate |
| CAS | 72590-77-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 H40 O7 |
| Quantity | 250 mg |
| Synonym | (11β)-11-Hydroxy-17-(1-oxobutoxy)-21-(1-oxopropoxy)-pregn-4-ene-3,20-dione, 17α-Butyryloxy-11β-hydroxy-21-propionyloxy-4-pregnene-3,20-dione, Hydrocortisone 17-butyrate 21-propionate, Hydrocortisone buteprate, Hydrocortisone probutate, Pandel, TS 408 |
| InChI Formula | InChI=1S/C28H40O7/c1-5-7-24(33)35-28(22(31)16-34-23(32)6-2)13-11-20-19-9-8-17-14-18(29)10-12-26(17,3)25(19)21(30)15-27(20,28)4/h14,19-21,25,30H,5-13,15-16H2,1-4H3/t19-,20-,21-,25+,26-,27-,28-/m0/s1 |
| SMILES | CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)CC |
| IUPAC Name | [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-10,13-dimethyl-3-oxo-17-(2-propanoyloxyacetyl)-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate |
| Molecular Weight (g/mol) | 488.61 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?