missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 17-Butyrate 21-Propionate, TRC

Product Code. 30393239
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393239 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393239 Supplier Toronto Research Chemicals Supplier No. TRCH71466550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocortisone 17-Butyrate 21-Propionate
CAS 72590-77-3
Purity Grade Notes HPLC
Molecular Formula C28 H40 O7
Quantity 50 mg
Synonym (11β)-11-Hydroxy-17-(1-oxobutoxy)-21-(1-oxopropoxy)-pregn-4-ene-3,20-dione, 17α-Butyryloxy-11β-hydroxy-21-propionyloxy-4-pregnene-3,20-dione, Hydrocortisone 17-butyrate 21-propionate, Hydrocortisone buteprate, Hydrocortisone probutate, Pandel, TS 408
InChI Formula InChI=1S/C28H40O7/c1-5-7-24(33)35-28(22(31)16-34-23(32)6-2)13-11-20-19-9-8-17-14-18(29)10-12-26(17,3)25(19)21(30)15-27(20,28)4/h14,19-21,25,30H,5-13,15-16H2,1-4H3/t19-,20-,21-,25+,26-,27-,28-/m0/s1
SMILES CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)CC
IUPAC Name [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-10,13-dimethyl-3-oxo-17-(2-propanoyloxyacetyl)-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate
Molecular Weight (g/mol) 488.61
Formula Weight 488.2774
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.