missing translation for 'onlineSavingsMsg'
Learn More

Hydrocinnamonitrile, 99+%

Product Code. 10429310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10429310 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10429310 Supplier Thermo Scientific Acros Supplier No. 173990100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 645-59-0 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 645-59-0
Melting Point -2°C to 1°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 261°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C9H9N
Linear Formula C6H5CH2CH2CN
Refractive Index 1.5210 to 1.523
MDL Number MFCD00001961
Quantity 10 g
Beilstein 09, 512
Specific Gravity 1
Synonym 3-phenylpropionitrile, benzenepropanenitrile, hydrocinnamonitrile, 2-phenylethyl cyanide, benzenepropionitrile, 2-cyanoethyl benzene, 3-phenylpropiononitrile, phenylpropionitrile, phenethyl cyanide, hydrocinnamique nitrile
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol,ethyl ether,slightly in chloroform
InChI Key ACRWYXSKEHUQDB-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CCC#N
IUPAC Name 3-phenylpropanenitrile
Molecular Weight (g/mol) 131.18
PubChem CID 12581
ChEBI CHEBI:85426
Formula Weight 131.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Hydrocinnamonitrile, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Hydrocinnamonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.