missing translation for 'onlineSavingsMsg'
Learn More

Hydrocinnamic-d5 Acid (phenyl-d5), TRC

Product Code. 30394557
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394557 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394557 Supplier Toronto Research Chemicals Supplier No. TRCH714598250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Phenylpropanoic Acid D5 (phenyl-D5)
CAS 35845-62-6
Molecular Formula C9 D5 H5 O2
Quantity 250 mg
Synonym Benzene-d5-propanoic acid (9CI), 3-(Pentadeuterophenyl)propionic acid, Pentadeuterohydrocinnamic acid, 3-Phenylpropanoic acid-d5, 3-(Phenyl-d5)propanoic acid, β-Phenylpropionic-d5 acid, β-(Phenyl-d5)propionic acid, Hydrocinnamic-d5 acid, Hydrocinnamic acid-d5, Hydrocinnamic-d5 Acid (phenyl-d5), Benzenepropanoic-d5 acid
InChI Formula InChI=1S/C9H10O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,11)/i1D,2D,3D,4D,5D
SMILES [2H]c1c([2H])c([2H])c(CCC(=O)O)c([2H])c1[2H]
IUPAC Name 3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid
Molecular Weight (g/mol) 155.2053
Formula Weight 155.0995
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.