missing translation for 'onlineSavingsMsg'
Learn More

Hydrocinnamic Acid 99%, ACROS Organics™

Brand:  Acros Organics 159101000

Code : 29

Additional Details : CAS Number : 501-52-0 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10103500

  • £24.90 / 100g


EU Stock 5
Add to basket

Specifications

Chemical Identifiers

Hydrocinnamic acid
501-52-0
99%
C6H5CH2CH2CO2H
MFCD00002771
99%
99%
C9H10O2
150.18
3-Phenylpropionic acid

Specifications

Hydrocinnamic acid
1.07
Clear yellow to yellow-green
100g
501-52-0
Authentic
99%
C9H10O2
Glass bottle
MFCD00002771
15, 4822
>112°C
279.0° to 281.0°C
47.0° to 50.0°C
99%
1.0700g/mL
Liquid
99%
C6H5CH2CH2CO2H
150.18
09, 508
3-Phenylpropionic acid
Videos
SDS