missing translation for 'onlineSavingsMsg'
Learn More

Hydrochlorothiazide, MedChemExpress

Product Code. 16440848
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16440848

Brand: Medchem Express HYB0252

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hydrochlorothiazide (HCTZ), an orally active diuretic drug of the thiazide class, inhibits transforming TGF-β/Smad signaling pathway. Hydrochlorothiazide has direct vascular relaxant effects via opening of the calcium-activated potassium (KCA) channel. Hydrochlorothiazide improves cardiac function, reduces fibrosis and has antihypertensive effect.

Hydrochlorothiazide (HCTZ), an orally active diuretic drug of the thiazide class, inhibits transforming TGF-β/Smad signaling pathway. Hydrochlorothiazide has direct vascular relaxant effects via opening of the calcium-activated potassium (KCA) channel. Hydrochlorothiazide improves cardiac function, reduces fibrosis and has antihypertensive effect.

Specifications

Chemical Name or Material Hydrochlorothiazide
CAS 58-93-5
Color White
Purity Grade Notes Research
Molecular Formula C7H8ClN3O4S2
Quantity 5 g
Synonym HCTZ
Solubility Information DMSO : 50 mg/mL (167.93 mM; Need ultrasonic)
SMILES O=S(C1=C(Cl)C=C(C2=C1)NCNS2(=O)=O)(N)=O
Molecular Weight (g/mol) 297.74
Formula Weight 297.74
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.