missing translation for 'onlineSavingsMsg'
Learn More

Hydrochlorothiazide, MedChemExpress

Product Code. 16458438
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458438 10 g 10g
16440848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458438 Supplier Medchem Express Supplier No. HYB0252
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hydrochlorothiazide (HCTZ), an orally active diuretic drug of the thiazide class, inhibits transforming TGF-β/Smad signaling pathway. Hydrochlorothiazide has direct vascular relaxant effects via opening of the calcium-activated potassium (KCA) channel. Hydrochlorothiazide improves cardiac function, reduces fibrosis and has antihypertensive effect.

Hydrochlorothiazide (HCTZ), an orally active diuretic drug of the thiazide class, inhibits transforming TGF-β/Smad signaling pathway. Hydrochlorothiazide has direct vascular relaxant effects via opening of the calcium-activated potassium (KCA) channel. Hydrochlorothiazide improves cardiac function, reduces fibrosis and has antihypertensive effect.

Specifications

Chemical Name or Material Hydrochlorothiazide
CAS 58-93-5
Color White
Purity Grade Notes Research
Molecular Formula C7H8ClN3O4S2
Quantity 10 g
Synonym HCTZ
Solubility Information DMSO : 50 mg/mL (167.93 mM; Need ultrasonic)
SMILES O=S(C1=C(Cl)C=C(C2=C1)NCNS2(=O)=O)(N)=O
Molecular Weight (g/mol) 297.74
Formula Weight 297.74
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.