missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid 34-37%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical™
83.79 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
94.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "21663" to get your promotional price.
94.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Description
ClH, CAS Number-7647-01-0, 500mL, 57 deg.C, Colorless, 36.46g/mol, -35 deg.C, 11324, Liquid, 1.27, 125mbar at 20 deg.C, 1.8 mPaS at 15 deg.C
Specifications
pH | 1 |
Color | Colorless |
Melting Point | -35°C |
Boiling Point | 57°C |
Vapor Pressure | 125mbar at 20°C |
Quantity | 500 mL |
Viscosity | 1.8 mPaS at 15°C |
Physical Form | Liquid |
Formula Weight | 36.46g/mol |
Safety and Handling
Product Identifier
- Hydrochloric acid, 34-37%
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P353-Rinse skin with water/shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrochloric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'