missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, Volumetric, 1M HCl (1N), Solstice

Product Code. 15676840 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15676840 1 L Poly bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15676840 Supplier Honeywell-Fluka Supplier No. 717631L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 1 M HCl (1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

pH ∼1.0
Color Colorless
CAS 7647-01-0
Melting Point <0°C
Boiling Point >100°C (1013hPa)
Density ∽1.010g/cu.cm at 20°C
Odor Odorless
Packaging Poly bottle
Vapor Pressure 190mmHg (25°C)
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324
UN Number UN1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Completely miscible in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES [H+].[Cl-]
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid solution, Volumetric, 1M HCl (1N)
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.