missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, Volumetric, 0.1M HCl (0.1N), Honeywell™ Fluka™

Product Code. 15677510 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
10 L
Packaging:
Poly Bottle
Poly Pail
Unit Size:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15677510 1 L Poly Bottle 1 L
15667510 10 L Poly Pail 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15677510 Supplier Honeywell-Fluka Supplier No. 940151L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH Acidic
Color Colorless
Boiling Point 100°C
Density 1g/cm3 at 20°C
Odor Odorless
Packaging Poly Bottle
Vapor Pressure 190mmHg (25°C)
Quantity 1 L
Physical Form Liquid
UN Number UN1789
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid solution, Volumetric, 0.1M HCl (0.1N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.