missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid solution, 6M EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12990854 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12990854 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12990854 Supplier Fisher Chemical Supplier No. VL 021 1000
  • ONLINE EXCLUSIVE PRICE
    £91.75
    £57.86 / 1000mL
    Save £33.89
    36% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 7647-10-1
Molecular Formula Cl2Pd
Concentration 6M
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Pd++]
IUPAC Name dichloropalladium
Molecular Weight (g/mol) 177.32
PubChem CID 24290
Quantity 1000 mL
ChEBI CHEBI:53434
Grade Eu. Ph. Reference
Chemical Name or Material Hydrochloric Acid
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid 6M. Volumetric Solution European Pharmacopoeia Ref: 3001500
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.