missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 0.5 M HCl (0.5N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.5 M HCl (0.5N)
£20.34 - £219.46
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15644950
|
Honeywell-Fluka
35329-1L |
1 L |
£20.34
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664950
|
Honeywell-Fluka
35329-5L-VP |
5 L |
£125.48
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654950
|
Honeywell-Fluka
35329-5L |
5 L | |||||||
15674950
|
Honeywell-Fluka
35329-6X1L |
6 x 1 L |
£129.53
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15634950
|
Honeywell-Fluka
35329-10L-VP |
10 L |
£219.46
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624950
|
Honeywell-Fluka
35329-10L |
10 L | |||||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
7647-01-0 | |
190 mmHg (25°C) | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric acid solution |
0.5M | |
ClH | |
UN1789 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrochloric acid solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid