missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid solution, 0.05M, Solstice

Product Code. 15664930 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
500mL
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15664930 6 x 500 mL Pack of 6
15644930 500 mL 500mL
15634930 1 L 1 L
15654930 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15664930 Supplier Honeywell-Fluka Supplier No. 353206X500ML
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 0.05 M HCl (0.05N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Vapor Pressure 190 mmHg (25°C)
Quantity 6 x 500 mL
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46
Chemical Name or Material Hydrochloric acid solution, volumetric, 0.05 M HCl (0.05N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.