missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid/Nitric Acid Blank, VHG™
Description
Hydrochloric Acid/Nitric Acid Blank
Specifications
Specifications
| Chemical Name or Material | Hydrochloric Acid/Nitric acid |
| CAS | 7647-01-0, 7697-37-2 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Hydrochloric Acid/Nitric acid |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3/HCl |
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?