missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, For determination of mercury, Reag. ISO, Reag. Ph. Eur., fuming (Hg ≤0.0000005%), ≥37%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
4 x 2.5 L
Unit Size:
Pack of 4
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 42°C |
| CAS Min % | ≥50.0000%, ≥30.0000% |
| CAS Max % | <70.0000%, <50.0000% |
| Density | 1.2 g/mL at 25 °C(lit.) |
| Packaging | Poly Bottle |
| Residue after Ignition | ≤0.0005% (as SO4) |
| Vapor Pressure | 3.23psi (21.1°C), 7.93psi (37.7°C) |
| Quantity | 4 x 2.5 L |
| Linear Formula | HCl |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction