missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Diluted, NF, Macron Fine Chemicals™

Product Code. p-7138031
Click to view available options
Quantity:
2.5 L
19 L
Packaging:
Glass bottle
Poly pail
Unit Size:
19 L
2.5 L
This item is not returnable. View return policy

Product Code. 15518034

Brand: Macron Fine Chemicals 260845

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Conforms to the requirements of the National Formulary (NF).

  • Meets the requirements necessary for use in pharmaceutical processing and prescription compounding
  • Product is regulated by the Food and Drug Administration
  • Manufactured under current Good Manufacturing Practices (cGMP)

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance Clear
Color Colorless
Melting Point 0°C
Boiling Point 85°C
Concentration 9.5 to 10g/100mL
Density 1g/cm3
Heavy Metals (as Pb) Max. 0.0005%
Identification Passes test
Packaging Glass bottle
Residue after Ignition Max. 0.01% (≤2mg)
Vapor Pressure 23 hPa
Quantity 2.5 L
Physical Form Solution
Solubility Information Fully miscible in water
Formula Weight 36.46g/mol
Percent Purity 10%
Grade NF
Chemical Name or Material Hydrochloric Acid
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.