Learn More
Hydrochloric Acid Concentrated Volumetric Solution Ampule, Reagecon
Fully factorized, high purity, stable product, developed and tested for titrations.
Brand: Reagecon HC0201L
Code : NEW
Additional Details : CAS Number : 7647-01-0 Weight : 0.12000kg Transport : UN number : 1789 Chem class : 8 Pack group : II
Description
- Use to make 1 L of 0.2 M (0.2 N) Hydrochloric Acid
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- When the product is accurately diluted to one liter in a volumetric flask, it can then be used as an analytical volumetric solution, to a specification of ± 0.2%.
- The product must be diluted accurately to one liter, in a volumetric flask under controlled temperature conditions, for optimal performance.
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection of any of several properties.
Specifications
0.2 M (0.2 N) | |
Ampule | |
1 Ampule | |
Liquid |
Safety and Handling
- HYDROCHLORIC ACID TO MAKE ON DILUTION 1L OF 0.2N SOLUTION
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid 37%