missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate, For 1 L standard solution, 0.01 M HCl (0.01 N), Honeywell Fluka™

Product Code. 15685260 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15685260 6 x 1 Ea. Pack of 6
15675260 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15685260 Supplier Honeywell-Fluka Supplier No. 382726X1EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 1 L standard solution, 0.01 M HCl (0.01 N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Boiling Point 100°C
CAS Min % ≥99.0000%
Concentration 0.01 M HCl (0.01 N)
Density 1.0 g/cm3
CAS Max % <1.0000%
Packaging Ampoule
Vapor Pressure 190 mmHg (25°C)
Quantity 6 x 1 Ea.
Linear Formula HCl
Physical Form Concentrate
UN Number UN1789
IUPAC Name chlorane
Formula Weight 36.5
Chemical Name or Material Hydrochloric acid concentrate, For 1 L standard solution, 0.01 M HCl (0.01 N)
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid concentrate
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.