missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT™, J.T. Baker™

Product Code. 10210051 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
50 mL
150 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10210051 50 mL Each
10107860 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10210051 Supplier JT Baker Supplier No. 4655

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Laboratory, Research, or Manufacturing Use

  • Management of Change (MOC) category = N
  • (1/10 equiv. = 3.646 g HCl)
  • Makes 0.1 N solution after dilution to 1000 mL

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Concentration 0.1 mol/L
Density 1 g/cm³ (20°C)
Packaging DILUT-IT™ Ampule (LDPE)
Normality Passes Test
Quantity 50 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.