missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Concentrate, 0.1N, Honeywell™ Fluka™

Product Code. 15657070 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Poly bottle
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15657070 250 mL Poly bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15657070 Supplier Honeywell-Fluka Supplier No. 77528250ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

pH >0.1 (at 20°C)
Color Colorless
CAS 7647-01-0
Density ∽1.072g/cu.cm at 20°C
Odor Stinging
Packaging Poly bottle
Vapor Pressure 190mmHg (25°C)
Quantity 250 mL
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
UN Number UN1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Completely miscible in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid concentrate, 0.1N, Dissolution Media Concentrate, Dilute to 10L to conform to USP and EP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.