missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Blank, VHG™
Description
Hydrochloric Acid Blank
Specifications
Specifications
| Chemical Name or Material | Hydrochloric Acid |
| CAS | 7647-01-0 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Hydrochloric Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | Cl H |
| Synonym | Hydrochloric Acid, Hydrogen Chloride, Muriatic Acid, Chlorohydric Acid |
| InChI Formula | InChI=1S/ClH/h1H |
| SMILES | Cl |
| IUPAC Name | chlorane |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?